HPLC Analysis of Propoxyphene and Methadone on Ascentis® Phenyl
HPLC Analysis of Propoxyphene and Methadone on Ascentis® Phenyl
Conditions
column Ascentis Phenyl, 15 cm x 4.6 mm I.D., 5 µm particles (581616-U)
column temp. 35 C
mobile phase [A] water with 0.1% ammonium acetate: [B] methanol; (20:80, A:B)
flow rate 1.0 ml/min
sample 50 µg/mL in 20:80, water:methanol
injection 10 µl
detector UV, 220 nm
Description
Categories Analytical Chromatography, Forensics and Toxicology, Pharmaceutical Analysis, drugs of abuse, analgesics, Analgesics
Featured Industry Forensics and Toxicology