HPLC Analysis of Protein Standards on Zenix® SEC-300 using Different Buffer Systems
HPLC Analysis of Protein Standards on Zenix® SEC-300 using Different Buffer Systems
Conditions
column Zenix SEC-300, 30 cm x 4.6 mm I.D., 3 µm particles (Z777028)
column temp. 25 °C
mobile phase [A] 0.15 M potassium phosphate monobasic, pH 7.0 with phosphoric acid; [B] 0.1 M ammonium bicarbonate (pH unadjusted)
flow rate 0.3 mL/min
sample in mobile phase
injection 2.5 µL
detector 1000 psi
detector UV, 280 nm
Description
Analysis Note This application compares selectivity of proteins when chromotographed by size exclusion with different buffer systems.
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Zenix is a registered trademark of Sepax Technologies