HPLC Analysis of Proteins (2) on BIOshell™ A400 Protein C4
HPLC Analysis of Proteins (2) on BIOshell™ A400 Protein C4
Conditions
column BIOshell A400 Protein C4, 10 cm X 2.1 mm I.D., 3.4 µm particles (66825-U)
column temp. 30 °C
mobile phase [A] 75:25, (0.1% TFA in water):(0.1% TFA in acetonitrile) ; [B] 25:75, (0.1% TFA in water):(0.1% TFA in acetonitrile)
gradient 15 to 75% B in 10 min
flow rate 0.3 mL/min
pressure 2410 psi (166 bar)
sample protein mixture, g/L ea: superoxide dismutase, 0.3; bovine serum albumin, 0.3; alpha-lactalbumin, 0.2; beta-lactoglobulin A, 0.3; catalase, 0.3; (carbonic anhydrase, 0.3; beta-Amylase, 1.0; glucose-6-phosphate dehydrogenase, 1.0; thioredoxin, 0.3; phosphorylase b, 2.5.)
injection 1 µL
detector UV, 215 nm
Description
Analysis Note This application demonstrates the utility of the BIOshell A400 Protein C4 column for the reversed-phase chromatography of proteins from a variety of sources and of different molecular weights.
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information BIOshell is a trademark of Sigma-Aldrich Co. LLC