Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Proteins on TSKgel® DEAE-NP
HPLC Analysis of Proteins on TSKgel® DEAE-NP
Conditions
column TSKgel DEAE-NPR, 3.5 cm x 4.6 mm I.D., 2.5 µm particles (813075)
mobile phase [A] 20 mM Tris-HCl buffer, pH 8.0; [B] 0.5 M sodium chloride in A
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation