Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Proteins on Mono Q HR 5/5
HPLC Analysis of Proteins on Mono Q HR 5/5
Conditions
column Mono Q HR 5/5, 10 µm particles (54807)
mobile phase [A]: 20 mM bis-Tris, pH 6.0; [B]: A + 1M sodium chlorde
flow rate 1.0 ml/min
sample human placental lactogen fraction from size exclusion medium
injection 2.5 mL
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins