Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Proteins and Peptides on Proteomix® WCX-NP1.7 Demonstrating Lot-to-Lot Reproducibility
HPLC Analysis of Proteins and Peptides on Proteomix® WCX-NP1.7 Demonstrating Lot-to-Lot Reproducibility
Conditions
column Proteomix WCX-NP1.7, 5 cm x 4.6 mm I.D., 1.7 µm particles (Z777180)
column temp. ambient
mobile phase (A) 20 mM sodium phosphate, pH 6.5; (B) A + 1.0 M NaCl
gradient 0 to 100% B in 25 min, 15 min prewash
flow rate 0.5 ml/min
sample 1 mg/mL
injection 5 µl
detector UV, 214 nm
Description
Categories Analytical Chromatography, Proteins, Peptides
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Proteomix is a registered trademark of Sepax Technologies