Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Proteins by Size Exclusion on Zenix® Columns, Effect of Pore Size
HPLC Analysis of Proteins by Size Exclusion on Zenix® Columns, Effect of Pore Size
Conditions
column Zenix SEC-150, 30 cm x 7.8 mm I.D., 3 µm, 150 Angstrom particles (Z777018)
column Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 µm, 300 Angstrom particles (Z777033)
column temp. ambient
mobile phase 150 mM sodium phosphate, pH 7
flow rate 1 ml/min
sample each 1 mg/mL except uracil, 2.5 mg/mL
injection 10 µl
detector UV, 214 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Zenix is a registered trademark of Sepax Technologies