HPLC Analysis of Proteins Comparing Ion Selectivity on Proteomix® SCX-NP5
HPLC Analysis of Proteins Comparing Ion Selectivity on Proteomix® SCX-NP5
Conditions
column Proteomix SCX-NP5, 5 cm x 4.6 mm I.D., 5 µm particles (Z777159)
column temp. 25 C
mobile phase [A] 10 mM potassium phosphate, dibasic, pH 7 with phosphoric acid [B] 0.3 M potassium potassium phosphate, pH 7 with phosphoric acid; ([C] 10 mM sodium phosphate, dibasic, pH 7 with phosphoric acid [D] 0.3 M sodium potassium phosphate, dibasic, pH 7 with phosphoric acid)
gradient 100% A to 100% B (or 100% C to 100% D) in 20 min
flow rate 0.5 ml/min
pressure 650 psitotal pressure (45 bar)
sample g/L ea: ribonuclease a, 0.38; cytochrome c, 0.15; lysozyme, 0.23; a-chymptrypsinogen, 0.23.
injection 5 µl
detector UV, 280 nm
Description
Analysis Note The choice of exchangable counterion can affect retention and selectivity when performing ion exchange chromatography of proteins.
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Proteomix is a registered trademark of Sepax Technologies