HPLC Analysis of Proteins on BIOshell™ A160 Peptide C18
HPLC Analysis of Proteins on BIOshell™ A160 Peptide C18
Conditions
column BIOshell A160 Peptide C18, 15 cm x 2.1 mm I.D., 2.7 µm particles (66905-U)
column temp. 35 °C
mobile phase [A] 75:25, 0.1% TFA in water: 0.08% TFA in acetonitrile; [B] 25:75, 0.1% TFA in water: 0.08% TFA in acetonitrile
gradient held at 0% B for 0.5 min; 0 to 100% B in 7 min
flow rate 0.3 mL/min
pressure 4743 psi (327 bar)
sample 0.2 - 0.6 g/L in mobile phase A
injection 1 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information BIOshell is a trademark of Sigma-Aldrich Co. LLC