HPLC Analysis of Proteins on SRT® SEC-300, 30 cm x 7.8 mm I.D., 5 μm particles
HPLC Analysis of Proteins on SRT® SEC-300, 30 cm x 7.8 mm I.D., 5 μm particles
Conditions
column SRT® SEC-300, 30 cm x 7.8 mm I.D., 5 µm particles (Z777051)
column temp. 25 °C
mobile phase 150 mM potassium phosphate, pH 7.0
flow rate 1.0 mL/min
sample 1-10 g/L in buffer
injection 10 µL
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information SRT is a registered trademark of Sepax Technologies