Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Proteins on SRT® SEC-300, 30 cm x 7.8 mm I.D., 5 μm
HPLC Analysis of Proteins on SRT® SEC-300, 30 cm x 7.8 mm I.D., 5 μm
Conditions
column SRT(R) SEC-300, 30 cm x 7.8 mm I.D., 5 µm particles (Z777059)
column temp. 25 C
mobile phase 150 mM potassium phosphate monobasic, pH 7.0
flow rate 1.0 ml/min
sample 1-10 g/L in buffer
injection 10 µl
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information SRT is a registered trademark of Sepax Technologies