HPLC Analysis of Proteins on SRT® SEC-500, 30 cm x 7.8 mm I.D.
HPLC Analysis of Proteins on SRT® SEC-500, 30 cm x 7.8 mm I.D.
Conditions
column SRT(R) SEC-500, 30 cm x 7.8 mm I.D., 5 µm particles (Z777057)
column temp. 25 C
mobile phase 150 mM potassium phosphate monobasic, pH 7.0
flow rate 1.0 ml/min
sample 1-10 g/L in buffer
injection 10 µl
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information SRT is a registered trademark of Sepax Technologies