HPLC Analysis of Proteins on Antibodix® WCX-NP10, Comparison of WCX Columns
HPLC Analysis of Proteins on Antibodix® WCX-NP10, Comparison of WCX Columns
Conditions
column Antibodix WCX-NP10, 25 cm x 4.6 mm I.D., 10 µm particles (Z777272)
column temp. ambient
mobile phase (A) 10 mM sodium phosphate buffer, pH 6.0; (B) A + 1.0 M NaCl
gradient 0 to 100% B in 42 min
flow rate 1.0 ml/min
sample 1 mg/mL each
injection 5 µl
detector UV, 214 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Antibodix is a registered trademark of Sepax Technologies