HPLC Analysis of Proteins on Proteomix® WCX-NP5
HPLC Analysis of Proteins on Proteomix® WCX-NP5
Conditions
column Proteomix WCX-NP5, 5 cm x 4.6 mm I.D., 5 µm particles (Z777193)
column temp. 25 °C
mobile phase [A] 10 mM sodium phosphate, dibasic, pH 7 with phosphoric acid; [B] 10 mM sodium phosphate, dibasic, 1M sodium chloride, pH 7 with phosphoric acid
gradient 0 to 60% in 20 min
flow rate 0.3 mL/min
pressure 420 psitotal pressure (29 bar)
sample ribonuclease a, 0.38; cytochrome c, 0.15; lysozyme, 0.23; a-chymptrypsinogen, 0.23 g/L
injection 5 µL
detector UV, 280 nm
Description
Analysis Note Four commonly studied proteins were chromatographed using a Proteomics WCX-NP5 Column to examine retention time and resolution. It was found that the Proteomics WCX-NP5 Column does not resolve the proteins as well as the Proteomics SCX-NP5, strong cation exchange column.
Featured Industry Life Science and Biopharma
Legal Information Proteomix is a registered trademark of Sepax Technologies