HPLC Analysis of Proteins with Different pI Values on Proteomix® SCX-NP3
HPLC Analysis of Proteins with Different pI Values on Proteomix® SCX-NP3
Conditions
column Proteomix SCX-NP3, 5 cm x 4.6 mm I.D., 3 µm particles (Z777151)
mobile phase (A) 10 mM sodium phosphate, pH 6.0; (B) A + 1 M NaCl
gradient 0 to 100% B in 15 min
flow rate 0.5 mL/min
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Proteomix is a registered trademark of Sepax Technologies