Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Purified Adeno-associated Virus (AAV) on TSKgel® SP-NPR
HPLC Analysis of Purified Adeno-associated Virus (AAV) on TSKgel® SP-NPR
Conditions
column TSKgel SP-NPR, 3.5 cm x 4.6 mm I.D., 2.5 µm particles (813076)
mobile phase [A] 50 mM HEPES, 1 mM EDTA, 5 mM MgCl, pH 7.5; [B] 50 mM HEPES, 1 mM EDTA, 5 mM MgCl, 0.5 M NaCl, pH 7.5
gradient 20 to 100% B in 10 column volumes
flow rate 1 ml/min
sample purified adeno-associated virus (AAV)
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.