HPLC Analysis of Pyridoxal (Vitamin B6) on SeQuant® ZIC-HILIC
HPLC Analysis of Pyridoxal (Vitamin B6) on SeQuant® ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 150 x 4.6 mm, 5 µm, 200 Ã… (1.50455.0001)
mobile phase [A] acetonitrile; [B] ammonium formate;; (90:10, A:B)
flow rate 0.5 ml/min
pressure 230 psi (1.6 MPa)
injection 10 µl
detector 254 nm
Description
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany