HPLC Analysis of Rabeprazole Enantiomers on Astec® CYCLOBOND® I 2000 DMP
HPLC Analysis of Rabeprazole Enantiomers on Astec® CYCLOBOND® I 2000 DMP
Conditions
column CYCLOBOND I 2000 DMP, 25 cm x 4.6 mm I.D., 5 µm particles (20724AST)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] methanol; [C] acetic acid; [D] ammonium hydroxide; (95:5:0.4:0.1, A:B:C:D)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 5 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Antiulcer Compounds, Pharmaceutical (small molecule), Antiulcer Drugs, All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC