HPLC Analysis of Ribonucleosides on Ascentis® Si
HPLC Analysis of Ribonucleosides on Ascentis® Si
Conditions
column Ascentis Si, 10 cm x 2.1 mm I.D., 5 µm particles (581508-U)
column temp. 35 °C
mobile phase [A] 150 mM ammonium formate (pH 3.0, with formic acid): [B] acetonitrile; (10:90, A:B)
flow rate 0.2 mL/min
sample as indicated in mobile phase
injection 1 µL
detector UV, 270 nm
Description
Categories Analytical Chromatography, Nucleic Acids