HPLC Analysis of Risedronate on SeQuant ZIC-HILIC
HPLC Analysis of Risedronate on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 100 x 4.6 mm, 5 µm, 200 Ã… (1.50453.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] ammonium acetate; (80:20, A:B)
flow rate 1.0 mL/min
injection 50 µL
detector 260 nm