HPLC Analysis of Ritalinic Acid and Ritalin (Methylphenidate) Enantiomers on Astec® CHIROBIOTIC® V2 with T2 Guard Column
HPLC Analysis of Ritalinic Acid and Ritalin (Methylphenidate) Enantiomers on Astec® CHIROBIOTIC® V2 with T2 Guard Column
Conditions
column CHIROBIOTIC V2, 15 cm x 4.6 mm I.D., 5 µm particles (15023AST)
column CHIROBIOTIC T2 guard column, 2 cm x 4 mm I.D., 5 µm particles (16100AST (requires holder 21150AST))
column temp. 25 °C
mobile phase [A] 20 mM ammonium acetate, pH 4.1; [B] methanol; (7:93, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 2 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Forensics and Toxicology, Chiral Applications, AD/HD Drugs, PsychostiµLants, ADHD drugs, Clinical, Forensic & Toxicology, psychostiµLant, Pharmaceutical (small molecule), Drugs of Abuse, All Pharmaceutical Applications, Therapeutic Drug Monitoring
Featured Industry Forensics and Toxicology
Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC