HPLC Analysis of Ritalinic Acid Enantiomers on Astec® CHIROBIOTIC® T2
HPLC Analysis of Ritalinic Acid Enantiomers on Astec® CHIROBIOTIC® T2
Conditions
column CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 µm particles (16024AST)
column temp. 25 °C
mobile phase [A] 10 mM ammonium acetate, pH 5.5; [B] acetonitrile; (70:30, A:B)
flow rate 0.9 mL/min
sample 1 mg/mL in methanol
injection 2 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Forensics and Toxicology, Chiral Applications, Clinical, Forensic & Toxicology, psychostiµLant, Pharmaceutical (small molecule), AD/HD Drugs, ADHD drugs, Therapeutic Drug Monitoring, Drugs of Abuse, All Pharmaceutical Applications
Featured Industry Forensics and Toxicology
Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC