Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of RNA on Proteomix® WAX-NP5
HPLC Analysis of RNA on Proteomix® WAX-NP5
Conditions
column Proteomix WAX-NP5, 15 cm x 4.6 mm I.D., 5 µm particles (Z777262)
column temp. Ambient
mobile phase (A) 20 mmol Tris, pH 6.0; (B): A + 1.0 M NaCl
gradient 20%-100%B (40 min)
flow rate 0.6 ml/min
pressure 1015 psi (70 bar)
sample RNA
injection 5 µl
detector UV, 260 nm
Description
Categories Analytical Chromatography, Nucleic Acids
Featured Industry Life Science and Biopharma
Legal Information Proteomix is a registered trademark of Sepax Technologies