HPLC Analysis of RP Peptide Performance Standard on Discovery® BIO Wide Pore C18, No TFA
HPLC Analysis of RP Peptide Performance Standard on Discovery® BIO Wide Pore C18, No TFA
Conditions
column Discovery BIO Wide Pore C18, 15 cm x 2.1 mm I.D., 3 µm particles (567202-U)
column temp. ambient
mobile phase [A] 25 mM formic acid in water; [B] (50:50) 25 mM formic acid in water:20 mM formic acid in acetonitrile
flow rate 0.20 ml/min
sample RP Peptide Performance Standard, p/n RPS-P0010 (Alberta Peptide Institute)
injection 1 µl
detector +ESI
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC