HPLC Analysis of Ruelene (Cruformate) Enantiomers on Astec® CYCLOBOND® I 2000
HPLC Analysis of Ruelene (Cruformate) Enantiomers on Astec® CYCLOBOND® I 2000
Conditions
column CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 µm particles (20024AST)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] acetic acid; [C] TEA; (100:0.3:0.2, A:B:C)
flow rate 0.6 mL/min
sample Ruelene (cruformate), 1 mg/mL in acetonitrile
injection 5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pesticides & Herbicides
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC