HPLC Analysis of Selegiline and Amphetamine Metabolites on Discovery® HS F5
HPLC Analysis of Selegiline and Amphetamine Metabolites on Discovery® HS F5
Conditions
column Discovery HS F5, 5 cm x 4.6 mm I.D., 5 µm particles (567513-U)
column temp. 40 °C
mobile phase [A] water (10 mM ammonium acetate, pH 4.0 with acetic acid: [B] acetonitrile (v/v), (25:75, A:B)
flow rate 1.0 mL/min
sample 50 µg/mL each in mobile phase
injection 10 µL
detector UV, 210 nm
Description
Categories Analytical Chromatography, Clinical, Forensics and Toxicology, drugs of abuse, antiparkinsonian, Amines
Featured Industry Clinical
Forensics and Toxicology
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC