HPLC Analysis of Simple Underivatized Sugars on apHera™ NH2
HPLC Analysis of Simple Underivatized Sugars on apHera™ NH2
Conditions
column apHera NH2, 15 cm x 4.6 mm I.D., 5 µm particles (56401AST)
column temp. 25 °C
mobile phase [A] water: [B] acetonitrile; (20:80, A:B)
flow rate 1.0 mL/min
sample 500 µg/mL in 30:70, water: acetonitrile
injection 10 µL
detector ELSD, 45 °C, 3.5 psi nitrogen
Description
Categories Analytical Chromatography, Carbohydrates
Featured Industry Food and Beverages
Legal Information apHera is a trademark of Sigma-Aldrich Co. LLC