Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Single Based Gun Surveillance Standard on Ascentis® Express Phenyl-Hexyl
HPLC Analysis of Single Based Gun Surveillance Standard on Ascentis® Express Phenyl-Hexyl
Conditions
column Ascentis Express Phenyl-Hexyl, 10 cm x 3.0 mm I.D., 2.0 µm particles (51617-U)
column temp. 30 C
mobile phase [A] water with 0.1% formic acid; [B] acetonitrile with 0.1% formic acid
gradient 60% B held for 0.8 min; to 75% B in 0.4 min; held at 75% B for 4 min
flow rate 0.4 ml/min
pressure 2620 psi (181 bar)
sample Single Based Gun Surveillance Standard (ERG-006), 50-200 µg/mL in acetonitrile, dilute 1:1 with water
injection 1 µl
detector UV, 250 nm
Description
Analysis Note Of interest to forensic analysis, this separation of gun shot residue compounds was achieved on an Ascentis Express Phenyl-Hexyl column using Cerilliant CRMs to provide reliable identification and quantification.
Categories Analytical Chromatography, Explosives
Featured Industry Forensics and Toxicology