HPLC Analysis of Six Enkephalin Peptides (Chiral and Achiral) on Astec® CHIROBIOTIC® T2
HPLC Analysis of Six Enkephalin Peptides (Chiral and Achiral) on Astec® CHIROBIOTIC® T2
Conditions
column CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 µm particles (16024AST)
mobile phase [A] 5 mM ammonium formate, pH 3.3; [B] acetonitrile; (25:75, A:B)
flow rate 0.5 ml/min
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Life Science & Biopharma, Peptides, All Biochemical Applications, Peptides & Proteins
Featured Industry Life Science and Biopharma
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC