HPLC Analysis of Steroidal Glycosides on Ascentis® Phenyl
HPLC Analysis of Steroidal Glycosides on Ascentis® Phenyl
Conditions
column Ascentis Phenyl, 15 cm x 4.6 mm I.D., 5 µm particles (581616-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium formate (pH 3 with formic acid); [B] methanol
flow rate 1.25 mL/min
sample as indicated in mobile phase A
injection 20 µL
detector ELSD at 45 °C, gain 6, 3.5 bar N2
Description
Categories Analytical Chromatography, Steroids
Featured Industry Vitamins, Nutraceuticals, and Natural Products