HPLC Analysis of Steroids on Ascentis® C18 (Mobile phase: 55:45)
HPLC Analysis of Steroids on Ascentis® C18 (Mobile phase: 55:45)
Conditions
column Ascentis C18, 15 cm x 4.6 mm I.D., 5 µm particles (581324-U)
column temp. 35 C
mobile phase [A] water; [B] acetonitrile; (55,45:A:B)
flow rate 1 ml/min
sample as indicated in 55:45, water:acetonitrile
injection 10 µl
detector UV, 220 nm
Description
Categories Analytical Chromatography, Clinical, Steroids, steroids
Featured Industry Clinical