HPLC Analysis of Steroids on Ascentis® C18 (Mobile phase: 60:40)
HPLC Analysis of Steroids on Ascentis® C18 (Mobile phase: 60:40)
Conditions
column Ascentis C18, 15 cm x 4.6 mm I.D., 5 µm particles (581324-U)
column temp. 35 °C
mobile phase [A] water; [B] acetonitrile; (60:40, A:B)
flow rate 1 mL/min
sample 50 µg/mL each in mobile phase
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Forensics and Toxicology, steroids, Steroids
Featured Industry Forensics and Toxicology