HPLC Analysis of Steroids on Ascentis® RP-Amide (Mobile phase: 55:45)
HPLC Analysis of Steroids on Ascentis® RP-Amide (Mobile phase: 55:45)
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 35 °C
mobile phase [A] water; [B] acetonitrile; (55:45, A:B)
flow rate 1 mL/min
sample as indicated in 55:45, water:acetonitrile
injection 10 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Clinical, Steroids, steroids
Featured Industry Clinical