HPLC Analysis of Steroids on Ascentis® Si
HPLC Analysis of Steroids on Ascentis® Si
Conditions
column Ascentis Si, 15 cm x 4.6 mm I.D., 5 µm particles (581512-U)
column temp. 35 °C
mobile phase [A] hexane; [B] ethanol; (88:12, A:B)
flow rate 1.0 mL/min
sample 50 µg/mL in 85:15, hexane:2-propanol
injection 10 µL
detector UV, 245 nm
Description
Categories Analytical Chromatography, Clinical, steroids, Steroids
Featured Industry Clinical
Vitamins, Nutraceuticals, and Natural Products