Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Steroids on Ascentis® Express F5, Acetonitrile Mobile Phase
HPLC Analysis of Steroids on Ascentis® Express F5, Acetonitrile Mobile Phase
Conditions
column Ascentis Express F5, 10 cm x 2.1 mm I.D., 2.7 µm particles (53569-U)
column temp. 35 C
mobile phase [A] water; [B] acetonitrile; (70:30, A:B)
flow rate 0.2 ml/min
sample 50 mg/L in 75:25, water:methanol
injection 2 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Clinical, steroids, Steroids
Featured Industry Clinical