HPLC Analysis of Steroids on Ascentis® Express F5 with Methanolic Mobile Phase
HPLC Analysis of Steroids on Ascentis® Express F5 with Methanolic Mobile Phase
Conditions
column Ascentis Express F5, 10 cm x 2.1 mm I.D., 2.7 µm particles (53569-U)
column temp. 35 C
mobile phase [A] water [B] methanol; (40:60, A:B)
flow rate 0.2 ml/min
sample 50 mg/L in 75:25, water:methanol
injection 0.2 ml/min
detector UV, 254 nm
Description
Categories Analytical Chromatography, Clinical, steroids, Steroids
Featured Industry Clinical