Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Steroids on Ascentis® Express F5
HPLC Analysis of Steroids on Ascentis® Express F5
Conditions
column Ascentis Express F5, 15 cm x 4.6 mm I.D., 2.7 µm particles (53591-U)
column temp. 35 C
mobile phase [A] water; [B] methanol; (50:50, A:B)
flow rate 0.8 ml/min
pressure 2393 psi (165 bar)
sample 10 µg/mL of each in 90: 10, water: methanol
injection 5 µl
detector UV, 240 nm
Description
Analysis Note Sample concentrations at 10 ug/mL in 90:10 water:methanol. Acetonitrile does not provide adequate selectivity on this phase. Mass spec was used for peak identification.
Categories Analytical Chromatography, Clinical, steroids, Steroids
Featured Industry Clinical