Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Steroids on Ascentis® Express C18
HPLC Analysis of Steroids on Ascentis® Express C18
Conditions
column Ascentis Express C18 10cm x 2.1mm, 2.7 µm particles (53823-U)
column temp. 40 C
mobile phase [A] water [B] acetonitrile (70:30, A:B)
flow rate 0.7 ml/min
pressure 5438 psi (375 bar)
sample 0.125 mg/mL each in 37.5/62.5 water/organic solvent
injection 2 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Steroids
Featured Industry Clinical
Pharmaceutical (small molecule)
General description This application demonstrates the suitability of Ascentis Express C18 for the efficient separation of 5 steroids.