HPLC Analysis of Steroids on Ascentis® Express Biphenyl
HPLC Analysis of Steroids on Ascentis® Express Biphenyl
Conditions
column Ascentis Express Biphenyl,  5 cm x 4.6 mm I.D., 2.7 µm particles (64059-U)
column temp. 30 °C
mobile phase [A] water; [B] acetonitrile
gradient 20 to 60% B in 6 min
flow rate 1.85 mL/min
pressure 4998 psi (344 bar)
sample 25 ng/mL each except 125 ng/mL deoxycorticosterone in 62.5:37.5, water:acetonitrile
injection 4 µL
detector UV, 215 nm
Description
Categories Analytical Chromatography, Steroids
Featured Industry Clinical