HPLC Analysis of Structural Isomers on Discovery® Zr-Carbon
HPLC Analysis of Structural Isomers on Discovery® Zr-Carbon
Conditions
column Discovery Zr-Carbon, 15 cm x 4.6 mm I.D., 3 µm particles (65730-U)
column temp. 60 °C
mobile phase [A] water; [B] acetonitrile; (20:80, A:B)
flow rate 0.5 mL/min
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Semivolatiles
Featured Industry Chemicals and Industrial Polymers
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC