HPLC Analysis of Substance P and Its Fragments on Discovery® C18
HPLC Analysis of Substance P and Its Fragments on Discovery® C18
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. 35 °C
mobile phase [A] water + 0.1% TFA; [B] acetonitrile + 0.1% TFA
gradient linear; 5-29%B (6 min); 29-34B (5 min)
flow rate 1 mL/min
sample Substance P in initial mobile phase
detector UV, 220 nm
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC