HPLC Analysis of Substituted Poly Aromatic Hydrocarbons using Ascentis® Express Biphenyl
HPLC Analysis of Substituted Poly Aromatic Hydrocarbons using Ascentis® Express Biphenyl
Conditions
column Ascentis Express Biphenyl, 10 cm x 2.1 mm, 2.7 µm particles (64065-U)
column temp. 35 C
mobile phase [A] Water; [B] Methanol; (30:70 A:B)
flow rate 0.5 ml/min
pressure 6200 psi (427 bar)
sample Uracil (10 µg/mL), Acenaphthenol (250 µg/mL), Acenaphthenone (20 µg/mL),   Acenaphthene (500 µg/mL), Prepared in 70:30 A:B
injection 1 µl
detector UV, 254 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express Biphenyl for the separation of substituted PAHs. It shows the suitability of this phase for separations of extended aromatics and their oxygenated counter parts. These compounds are potentional metabolites, used as synthetic intermediates, and in the study of dihydrodiol dehydrogenase activity in celluar systems.
Categories Analytical Chromatography, PAHs
Featured Industry Chemicals and Industrial Polymers
Environmental