HPLC Analysis of Sucrose on SeQuant® ZIC-HILIC with Refractive Index Detectio
HPLC Analysis of Sucrose on SeQuant® ZIC-HILIC with Refractive Index Detectio
Conditions
column SeQuant ZIC-HILIC, PEEK, 150 x 4.6 mm, 5 μm, 200 Å (1.50455.0001)
mobile phase [A] acetonitrile; [B] ammonium acetate; (80:20, A:B)
flow rate 1.0 mL/min
column temp. ambient
detector refractive index
injection: 20 μL
Description
Materials
Featured Industry Food and Beverages
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany