Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Sugars on SUPELCOSIL™ LC-NH2
HPLC Analysis of Sugars on SUPELCOSIL™ LC-NH2
Conditions
column SUPELCOSIL LC-NH2, 25 cm x 4.6 mm I.D., 5 µm particles (58338)
mobile phase [A] water; [B] acetonitrile; (25:75, A:B)
flow rate 1 ml/min
temp. ambient
sample 10 mg/mL in water
injection 2 µl
detector refractive index (RI)
Description
Categories Analytical Chromatography, Sweeteners, Carbohydrates
Featured Industry Food and Beverages
Legal Information SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC