HPLC Analysis of Sulconazole Enantiomers on Astec® Cellulose DMP
HPLC Analysis of Sulconazole Enantiomers on Astec® Cellulose DMP
Conditions
column Astec Cellulose DMP, 25 cm x 4.6 mm I.D., 5 µm particles (51099AST)
column temp. 30 C
mobile phase [A] heptane; [B] isopropanol; (80:20, A:B)
flow rate 1 ml/min
sample 1 g/L in 50:50, heptane:isopropanol
injection 15 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Antifungals
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC