Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Sulfa Drugs on Discovery® C18
HPLC Analysis of Sulfa Drugs on Discovery® C18
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. 20 C
mobile phase [A] methanol; [B] water with 1% acetic acid (20:80, A:B)
flow rate 1 ml/min
injection 10 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antibiotics
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC