HPLC Analysis of Sulfa Drugs on Titan™ C18
HPLC Analysis of Sulfa Drugs on Titan™ C18
Conditions
column Titan C18 5 cm x 2.1 mm I.D., 1.9 µm particle (577122-U)
column temp. 40 °C
mobile phase [A] water with 0.1% acetic acid; [B] acetonitrile with 0.1% acetic acid ; (90:10, A:B)
flow rate 0.4 mL/min
pressure 2670 psi (184 bar)
sample 25 µg/mL in 70:30, water:methanol
injection 2 µL
detector UV, 250 nm
Description
Categories Analytical Chromatography, Antibiotics, Antimicrobials
Featured Industry Clinical
Forensics and Toxicology
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC