HPLC Analysis of Sunscreens on Ascentis® C18
HPLC Analysis of Sunscreens on Ascentis® C18
Conditions
column Ascentis C18, 15 cm x 4.6 mm I.D., 5 µm particles (581324-U)
column temp. 35 °C
mobile phase [A] water, [B] methanol, (88:12, A:B)
flow rate 1 mL/min
sample 50 µg/mL in mobile phase
injection 10 µL
detector UV, 305 nm
Description
Categories Analytical Chromatography, Ketones, Esters
Featured Industry Cosmetics, Personal Care, and Cleaning Products