HPLC Analysis of Tamoxifen E/Z isomers and Related Compounds on Ascentis® Express RP-Amide
HPLC Analysis of Tamoxifen E/Z isomers and Related Compounds on Ascentis® Express RP-Amide
Conditions
column Ascentis Express RP-Amide, 10 cm x 3 mm I.D., 2.7 µm particles (53918-U)
column temp. 35 C
mobile phase [A] 0.1% formic acid in water; [B] 0.1% formic acid in acetonitrile
gradient held at 25% B for 2 min; 25 to 40% B in 15 min; 40 to 25% B in 0.1 min; held at 25% B for 2.9 min
flow rate 0.6 ml/min
pressure 2292 psi (158 bar)
sample 10 mg/L in 10:90, water: methanol
injection 2 µl
detector UV, 260 nm
Description
Categories Analytical Chromatography, Agonist/antagonist