HPLC Analysis of Taxols on Ascentis® Express F5
HPLC Analysis of Taxols on Ascentis® Express F5
Conditions
column Ascentis Express F5, 10 cm x 2.1 mm I.D., 2.7 µm particles (53569-U)
column temp. 30 °C
mobile phase [A] water; [B] acetonitrile; (60:40, A:B)
flow rate 0.3 mL/min
sample 25 mg/L in 70:30, water:methanol
injection 2 µL
detector UV, 227 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express F5 for the efficient separation of taxols.
Categories Analytical Chromatography, Antineoplastics